Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301060.1 | NP_001287989.1 | 9 - 10 | 1088 | 67 | |
NM_024848.2 | NP_079124.1 | 9 - 10 | 1088 | 67 | ||
XM_005244798.2 | XP_005244855.1 | 8 - 9 | 952 | 67 | ||
XM_011542169.1 | XP_011540471.1 | 9 - 10 | 1088 | 67 | ||
XM_011542170.2 | XP_011540472.1 | 9 - 10 | 1088 | 67 | ||
XM_011542172.2 | XP_011540474.1 | 8 - 9 | 952 | 67 | ||
XM_011542173.2 | XP_011540475.1 | 9 - 10 | 1088 | 67 | ||
XM_011542174.2 | XP_011540476.1 | 9 - 10 | 1088 | 67 | ||
XM_017002365.1 | XP_016857854.1 | 9 - 10 | 1088 | 67 | ||
GenBank mRNA | AK024003.1 | - | 9 - 10 | 891 | 67 | |
AK310323.1 | - | 8 - 9 | 952 | 67 | ||
BC021704.2 | - | 9 - 10 | 1044 | 4825983 | 67 | |
BU741473.1 | - | 2 - 3 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |