Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006035.3 | NP_006026.3 | 23 - 24 | 3302 | 128 | |
XM_005268227.1 | XP_005268284.1 | 24 - 25 | 3183 | 128 | ||
XM_005268230.4 | XP_005268287.1 | 24 - 25 | 3183 | 128 | ||
XM_011537387.1 | XP_011535689.1 | 24 - 25 | 3183 | 128 | ||
GenBank mRNA | AB032950.3 | - | 23 - 24 | 3293 | 128 | |
AF128625.1 | - | 23 - 24 | 3359 | 128 | ||
BC155541.1 | - | 23 - 24 | 3305 | 8860517 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons |