Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000467481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000467481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000467481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000050.hg.1 |
ENST00000360754.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000050.hg.1 |
ENST00000361479.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000050.hg.1 |
ENST00000414242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000050.hg.1 |
ENST00000538024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000050.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000050.hg.1 |
TC13000017.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000449056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000467481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000467481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000467481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
ENST00000496525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006474.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
ENST00000361479.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
ENST00000414242.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
NM_017520.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144247.hg.1 |
TC1300006474.hg.1 |
Clariom D TC |