Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
TC02002953.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
ENST00000402136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
ENST00000402545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
ENST00000407315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
ENST00000512346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047621.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000321679.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000497486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016366.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000402136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000402545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000407315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
ENST00000612200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
NM_001164356.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
NM_015963.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200220211.hg.1 |
TC0200016366.hg.1 |
Clariom D TC |