Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000571744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000571744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014307.hg.1 |
ENST00000338694.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014307.hg.1 |
ENST00000535086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014307.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014307.hg.1 |
TC17001069.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174173.hg.1 |
TC1700009600.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000338694.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000571744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000571744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000572291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000573966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
ENST00000575592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009600.hg.1 |
NM_053285.1 |
Refseq |