Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304472.1 | NP_001291401.1 | 12 - 13 | 1682 | 82 | |
NM_001304473.1 | NP_001291402.1 | 13 - 14 | 1708 | 82 | ||
NM_001304474.1 | NP_001291403.1 | 12 - 13 | 1653 | 82 | ||
NM_003741.3 | NP_003732.2 | 12 - 13 | 1682 | 82 | ||
NR_130747.1 | 10 - 11 | 1840 | 82 | |||
XM_011513254.1 | XP_011511556.1 | 12 - 13 | 1535 | 82 | ||
XM_011513256.1 | XP_011511558.1 | 10 - 11 | 1691 | 82 | ||
XM_017007388.1 | XP_016862877.1 | 11 - 12 | 1616 | 82 | ||
XM_017007389.1 | XP_016862878.1 | 11 - 12 | 1616 | 82 | ||
XM_017007390.1 | XP_016862879.1 | 11 - 12 | 1616 | 82 | ||
XM_017007391.1 | XP_016862880.1 | 11 - 12 | 1616 | 82 | ||
XM_017007392.1 | XP_016862881.1 | 12 - 13 | 1535 | 82 | ||
XM_017007393.1 | XP_016862882.1 | 11 - 12 | 1616 | 82 | ||
XM_017007394.1 | XP_016862883.1 | 10 - 11 | 1404 | 82 | ||
GenBank mRNA | AB208947.1 | - | 7 - 8 | 2233 | 82 | |
AF209928.1 | - | 12 - 13 | 1682 | 82 | ||
AF209930.1 | - | 13 - 14 | 1708 | 82 | ||
AF283325.1 | - | 12 - 13 | 1653 | 82 | ||
AK074147.1 | - | 10 - 11 | 1593 | 82 | ||
AK293456.1 | - | 12 - 13 | 1404 | 82 | ||
AK300213.1 | - | 4 - 5 | 1024 | 82 | ||
AK300476.1 | - | 1 - 2 | 111 | 82 | ||
AY358926.1 | - | 12 - 13 | 1572 | 82 | ||
BC112345.1 | - | 12 - 13 | 1467 | 40008417 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |