Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018358.2 | NP_060828.2 | 10 - 11 | 1163 | 92 | |
XM_011512959.2 | XP_011511261.1 | 10 - 11 | 1167 | 92 | ||
GenBank mRNA | AK002060.1 | - | 10 - 11 | 1140 | 92 | |
AK024758.1 | - | 4 - 5 | 362 | 92 | ||
AK092415.1 | - | 10 - 11 | 1046 | 92 | ||
AK097073.1 | - | 9 - 10 | 1430 | 92 | ||
AK290106.1 | - | 10 - 11 | 1035 | 92 | ||
AK299433.1 | - | 10 - 11 | 927 | 92 | ||
BC009253.2 | - | 10 - 11 | 1042 | 3028775 | 92 | |
BC035652.1 | - | 10 - 11 | 1037 | 92 | ||
BC051754.2 | - | 10 - 11 | 1037 | 6064628 | 92 | |
BC051884.1 | - | 10 - 11 | 1037 | 5105761 | 92 | |
KU178694.1 | - | 10 - 11 | 1030 | 92 | ||
KU178695.1 | - | 10 - 11 | 987 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |