Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001156.hg.1 |
TC01000072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001156.hg.1 |
ENST00000294600.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001156.hg.1 |
ENST00000462521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001156.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100145822.hg.1 |
TC0100006623.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000294600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
ENST00000462521.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006623.hg.1 |
NM_152492.2 |
Refseq |