Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013284.hg.1 |
ENST00000302451.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013284.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013284.hg.1 |
TC16000925.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600173945.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173945.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173945.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173945.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173945.hg.1 |
TC1600009656.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |