Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002060.hg.1 |
TC03000096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002060.hg.1 |
ENST00000396842.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002060.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000617675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
ENST00000617675.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000449565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000617675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
ENST00000617675.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006714.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146844.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146844.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146844.hg.1 |
ENST00000396842.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146844.hg.1 |
NM_033083.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146844.hg.1 |
TC0300006714.hg.1 |
Clariom D TC |