Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019446.hg.1 |
TC09001543.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019446.hg.1 |
ENST00000373840.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019446.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177756.hg.1 |
TC0900011396.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000373840.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
ENST00000451303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011396.hg.1 |
NM_016322.3 |
Refseq |