Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012247.hg.1 |
ENST00000290231.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012247.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012247.hg.1 |
TC20000896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012250.hg.1 |
ENST00000290231.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012250.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012250.hg.1 |
TC20000896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012248.hg.1 |
ENST00000290231.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012248.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012248.hg.1 |
TC20000896.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000164824.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164824.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164824.hg.1 |
TC2000009259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164824.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164824.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
ENST00000372291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009259.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164826.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164826.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164826.hg.1 |
TC2000009259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164826.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164826.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164825.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164825.hg.1 |
NM_020967.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164825.hg.1 |
TC2000009259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164825.hg.1 |
ENST00000290231.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164825.hg.1 |
NM_020967.2 |
Refseq |