Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133180.2 | NP_573441.2 | 4 - 5 | 226 | 112 | |
XM_005259020.1 | XP_005259077.1 | 4 - 5 | 226 | 112 | ||
XM_011527051.2 | XP_011525353.1 | 4 - 5 | 226 | 112 | ||
XM_011527052.2 | XP_011525354.1 | 4 - 5 | 226 | 112 | ||
GenBank mRNA | AF370395.1 | - | 4 - 5 | 178 | 112 | |
AK309305.1 | - | 4 - 5 | 226 | 112 | ||
AY074928.1 | - | 3 - 4 | 179 | 112 | ||
DA452167.1 | - | 4 - 5 | 226 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |