Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014981.1 | NP_055796.1 | 34 - 35 | 4854 | 99 | |
XM_011512559.2 | XP_011510861.1 | 35 - 36 | 4975 | 99 | ||
XM_017005922.1 | XP_016861411.1 | 25 - 26 | 4119 | 99 | ||
GenBank mRNA | AB023217.3 | - | 34 - 35 | 4854 | 99 | |
AK055436.1 | - | 11 - 12 | 1940 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |