Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2ML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282424.2 | NP_001269353.1 | 16 - 17 | 103 | ||
NM_144670.5 | NP_653271.2 | 27 - 28 | 103 | |||
XM_011520566.2 | XP_011518868.1 | 27 - 28 | 103 | |||
XM_011520567.2 | XP_011518869.1 | 27 - 28 | 103 | |||
XM_017018868.1 | XP_016874357.1 | 27 - 28 | 103 | |||
XM_017018869.1 | XP_016874358.1 | 27 - 28 | 103 | |||
XM_017018870.1 | XP_016874359.1 | 27 - 28 | 103 | |||
GenBank mRNA | AK096448.1 | - | 5 - 6 | 103 | ||
AK122624.1 | - | 21 - 22 | 103 | |||
AK123591.1 | - | 10 - 11 | 103 | |||
AK123592.1 | - | 16 - 17 | 103 | |||
AK123601.1 | - | 24 - 25 | 103 | |||
AK290355.1 | - | 27 - 28 | 103 | |||
AK302555.1 | - | 16 - 17 | 103 | |||
AL832139.1 | - | 27 - 28 | 103 | |||
AL832750.1 | - | 4 - 5 | 103 | |||
AL833479.1 | - | 25 - 26 | 103 | |||
BF354222.1 | - | 2 - 3 | 287 | 103 | ||
BX647674.1 | - | 9 - 10 | 103 | |||
BX648231.1 | - | 28 - 29 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |