Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013497.hg.1 |
ENST00000411493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006502.hg.1 |
TC04000454.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006502.hg.1 |
ENST00000273920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006502.hg.1 |
ENST00000456931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006502.hg.1 |
ENST00000509635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04006502.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154895.hg.1 |
TC0400007978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013497.hg.1 |
ENST00000411493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000505846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007978.hg.1 |
NR_120457.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000610545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154896.hg.1 |
TC0400007978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
ENST00000273920.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
ENST00000509635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
ENST00000618400.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
NM_001292017.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
NM_021204.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154894.hg.1 |
TC0400007978.hg.1 |
Clariom D TC |