Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01003293_m1
See other EYA1 GE Assays ›
Gene Symbol:
EYA1
Gene Name:
EYA transcriptional coactivator and phosphatase 1
Gene Aliases:
BOP, BOR, BOS1, OFC1
Chromosome Location:
Chr.8: 71197433 - 71548130 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01003293_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: EYA1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_000503.5
NP_000494.2
3 - 4
762
83
NM_001288574.1
NP_001275503.1
3 - 4
762
83
NM_001288575.1
NP_001275504.1
3 - 4
762
83
NM_172058.3
NP_742055.1
2 - 3
354
83
NM_172059.3
NP_742056.1
2 - 3
176
83
XM_011517483.2
XP_011515785.1
2 - 3
5062
83
XM_011517484.2
XP_011515786.2
4 - 5
520
83
XM_017013201.1
XP_016868690.1
4 - 5
548
83
XM_017013202.1
XP_016868691.1
4 - 5
392
83
XM_017013203.1
XP_016868692.1
4 - 5
520
83
XM_017013204.1
XP_016868693.1
4 - 5
521
83
XM_017013205.1
XP_016868694.1
4 - 5
520
83
XM_017013206.1
XP_016868695.1
5 - 6
501
83
XM_017013207.1
XP_016868696.1
4 - 5
519
83
XM_017013208.1
XP_016868697.1
3 - 4
787
83
XM_017013209.1
XP_016868698.1
4 - 5
409
83
XM_017013210.1
XP_016868699.1
4 - 5
521
83
XM_017013213.1
XP_016868702.1
2 - 3
5264
83
GenBank mRNA
AF467247.1
-
2 - 3
151
83
AJ000097.1
-
2 - 3
299
83
AJ000098.1
-
3 - 4
760
83
AK127798.1
-
3 - 4
262
83
AK295897.1
-
3 - 4
271
83
AK307379.1
-
3 - 4
589
83
BC121798.1
-
2 - 3
266
40084971
83
BC121799.1
-
2 - 3
266
40084967
83
DC360066.1
-
2 - 3
354
83
DC409057.1
-
2 - 3
409
83
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the AK127798.1, BC121798.1, NM_001288575.1, XM_017013213.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene EYA1
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
metanephros development
branching involved in ureteric bud morphogenesis
outflow tract morphogenesis
double-strand break repair
transcription, DNA-templated
pattern specification process
mesodermal cell fate specification
sensory perception of sound
anatomical structure morphogenesis
response to ionizing radiation
striated muscle tissue development
histone dephosphorylation
protein sumoylation
cellular protein localization
establishment or maintenance of apical/basal cell polarity
peptidyl-tyrosine dephosphorylation
aorta morphogenesis
outer ear morphogenesis
middle ear morphogenesis
regulation of neuron differentiation
positive regulation of DNA repair
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
neuron fate specification
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
positive regulation of epithelial cell proliferation
pharyngeal system development
lung epithelial cell differentiation
otic vesicle morphogenesis
positive regulation of secondary heart field cardioblast proliferation
cochlea morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA binding
protein tyrosine phosphatase activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
TC08001316.hg.1
HTA TC
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
ENST00000388740.3
Ensembl
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
ENST00000388742.4
Ensembl
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
ENST00000388743.2
Ensembl
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
NM_172058.3
Refseq
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
NM_172059.3
Refseq
Affymetrix Human Transcriptome Array 2.0
PSR08016975.hg.1
NM_172060.3
Refseq
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0