Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
TC04000952.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
ENST00000322224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
ENST00000347950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
ENST00000404286.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
ENST00000503156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
ENST00000515118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012800.hg.1 |
NM_032219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000322224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000347950.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000404286.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000503156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000507165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000512400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000515118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
NM_001294341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
NM_001294342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
NM_032219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000322224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000347950.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000404286.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000503156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000507165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000512400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
ENST00000515118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
NM_001294341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
NM_001294342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
NM_032219.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166315.hg.1 |
TC0400009695.hg.1 |
Clariom D TC |