Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014045.hg.1 |
TC05000969.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014045.hg.1 |
ENST00000303177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014045.hg.1 |
ENST00000521585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014045.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014041.hg.1 |
TC05000969.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014041.hg.1 |
ENST00000303177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014041.hg.1 |
ENST00000521585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014041.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014047.hg.1 |
TC05000969.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014047.hg.1 |
ENST00000303177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014047.hg.1 |
ENST00000521585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014047.hg.1 |
ENST00000521959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05014047.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
ENST00000521959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013266.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194654.hg.1 |
TC0500013266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194651.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194651.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194651.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194651.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194651.hg.1 |
TC0500013266.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000303177.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000517587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000517902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000519717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000519867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
ENST00000521278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
NM_015980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194659.hg.1 |
TC0500013266.hg.1 |
Clariom D TC |