Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000355531.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000388970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000418696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421164.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000421780.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000423742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000427658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000427658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000427658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000427658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000427658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000429663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000429663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000429663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000429663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000431757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000431757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000431757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000431757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000431757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000436447.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000437774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000450026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000450026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000450026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000450026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000454997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000457926.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000458301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000458301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000458301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000458301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000458301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
ENST00000557444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003226.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003227.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008908.hg.1 |
NR_003228.1 |
Refseq |