Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004039.hg.1 |
TC02000271.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004039.hg.1 |
ENST00000260605.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004039.hg.1 |
ENST00000443170.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004039.hg.1 |
ENST00000489222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004039.hg.1 |
NM_001193464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004039.hg.1 |
NM_016008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000260605.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000378587.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000462426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000479242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000489222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000605786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
NM_001193464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
NM_016008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000260605.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000378587.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000462426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000479242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000489222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
ENST00000605786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
NM_001193464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
NM_016008.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151252.hg.1 |
TC0200007401.hg.1 |
Clariom D TC |