Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017176.hg.1 |
TC01001090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017176.hg.1 |
ENST00000369317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017176.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017175.hg.1 |
TC01001090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017175.hg.1 |
ENST00000369317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017175.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216679.hg.1 |
TC0100015598.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000425134.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000486597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000488537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
ENST00000582401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015598.hg.1 |
NM_006472.5 |
Refseq |