Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902039 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902007 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902038 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902013 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329266 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329278 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329246 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329272 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902000 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16984058 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329248 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16984054 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329245 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902006 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329254 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902018 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902021 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329240 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329247 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329262 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30901994 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902022 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902012 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16984055 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902008 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30901987 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16984051 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30901981 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329242 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329269 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902041 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329281 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329271 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329274 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329231 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329261 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329235 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329273 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16984049 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329265 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30901996 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329233 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30902016 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P10329268 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
130068 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
154187 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
10885 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
940930 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
879999 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
849521 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
948966 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
33505 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
122934 |
AK000091.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
122934 |
AK172764.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
122934 |
AK222636.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
122934 |
BC011553.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
122934 |
CR602598.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
122934 |
NM_017659.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
164813 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
230371 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P395621 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P395621 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P129996 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
1526 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
16269 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P132633 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P132633 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
58182 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P57541947 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P57541948 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P03247273 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P03247275 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P395621 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_24_P395621 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901996 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902016 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902012 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902021 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902039 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984049 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902041 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901987 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902022 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902038 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329231 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329240 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329271 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902018 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901990 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901994 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329238 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901981 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902000 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329245 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902013 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902008 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984051 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329246 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329268 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329254 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329242 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329247 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329269 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984058 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329278 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902004 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984054 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329274 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329272 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329248 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329233 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902006 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901994 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329269 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329248 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902004 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902018 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329238 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902021 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329254 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329278 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902039 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901981 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984051 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902012 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902041 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902000 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984058 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902013 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984049 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902038 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902022 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329272 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329231 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902006 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329268 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329246 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901987 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329240 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329274 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329247 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901990 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329266 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16984054 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329233 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902008 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30902016 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329245 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329271 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30901996 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P10329242 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_24_P395621 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P129996 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P40822020 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P132633 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329239 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329271 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329273 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329276 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329230 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329278 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329251 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329280 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329236 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329238 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329252 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329241 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329274 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329245 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329248 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329269 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329244 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329272 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329247 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329255 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329268 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P10329234 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984046 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984045 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P10329229 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984048 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P10329230 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984044 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P10329228 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984043 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984047 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16984042 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P40822020 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P20651432 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P132633 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P03247266 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P132633 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P129996 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P395621 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
176938 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
10367 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
76109 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
14405 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P40822020 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20651432 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03247266 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P132633 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20651416 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03247272 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329238 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329273 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16984051 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329314 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329271 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329266 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329295 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329317 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329268 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329293 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329263 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329319 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329256 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329241 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329247 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16984049 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329272 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329284 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329236 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329258 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329306 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329288 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329245 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329274 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329230 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329301 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329278 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329255 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329285 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329251 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329243 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16984048 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16984042 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329234 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16984045 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329269 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329312 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329302 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329309 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329298 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329296 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329248 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329310 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329287 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329260 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329305 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329304 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16984054 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329283 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P10329291 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P132633 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P40822020 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03247266 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P20651432 |
- |
- |