Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001344.2 | NP_001001344.1 | 19 - 20 | 3475 | 121 | |
XM_005274690.3 | XP_005274747.1 | 21 - 22 | 3793 | 121 | ||
XM_005274691.3 | XP_005274748.1 | 20 - 21 | 3751 | 121 | ||
GenBank mRNA | BC130009.1 | - | 19 - 20 | 3408 | 40067815 | 121 |
U60414.1 | - | 19 - 20 | 3475 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |