Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000465258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000465258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000465258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000614001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000614001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
ENST00000614001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006316.hg.1 |
ENST00000357137.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006316.hg.1 |
ENST00000414396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006316.hg.1 |
ENST00000541156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006316.hg.1 |
ENST00000544094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006316.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006316.hg.1 |
TC22000327.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200169086.hg.1 |
TC2200009277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
TC2200009277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200169085.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000357137.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000417688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000428799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000465258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000465258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000465258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000482652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000614001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000614001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
ENST00000614001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_001204827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009277.hg.1 |
NM_022098.3 |
Refseq |