Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK000088.1 | - | 1 | 1230 | 159 | |
AK225183.1 | - | 1 | 1230 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |