Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003941.3 | NP_003932.3 | 4 - 5 | 771 | 79 | |
GenBank mRNA | AK289795.1 | - | 4 - 5 | 684 | 79 | |
AM295156.1 | - | 4 - 5 | 434 | 79 | ||
BC052955.1 | - | 4 - 5 | 721 | 5264663 | 79 | |
CR749438.1 | - | 5 - 6 | 676 | 79 | ||
D88460.1 | - | 4 - 5 | 688 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |