Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
TC12003212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
ENST00000261733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
ENST00000416293.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
ENST00000551906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
ENST00000552234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
ENST00000553044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011475.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000551450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
ENST00000551450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201644.hg.1 |
TC1200012707.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000261733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000416293.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000548536.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000549106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000551450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
ENST00000551450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_000690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012707.hg.1 |
NM_001204889.1 |
Refseq |