Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014258.hg.1 |
ENST00000245663.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014258.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014258.hg.1 |
ENST00000395104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014258.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014258.hg.1 |
TC20001039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168693.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |