Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005266307.3 | XP_005266364.1 | 16 - 17 | 4771 | 104 | |
XM_005266311.3 | XP_005266368.1 | 15 - 16 | 4687 | 104 | ||
XM_017020481.1 | XP_016875970.1 | 16 - 17 | 4712 | 104 | ||
XM_017020482.1 | XP_016875971.1 | 15 - 16 | 4387 | 104 | ||
GenBank mRNA | AB020660.1 | - | 4 - 5 | 2219 | 104 | |
AK074924.1 | - | 2 - 3 | 1447 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |