Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004423.hg.1 |
ENST00000360428.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004423.hg.1 |
ENST00000434452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004423.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004423.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004423.hg.1 |
TC18000435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
TC1800008385.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157118.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000360428.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000434452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
ENST00000584980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_001941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008385.hg.1 |
NM_024423.3 |
Refseq |