Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010269.hg.1 |
TC04000732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010269.hg.1 |
ENST00000296582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010269.hg.1 |
ENST00000508208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010269.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000505999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000506826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008947.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
ENST00000296582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
ENST00000508208.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
NM_018241.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400161462.hg.1 |
TC0400008947.hg.1 |
Clariom D TC |