Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001697.hg.1 |
TC09000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001697.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001697.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147544.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147544.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147544.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147544.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147544.hg.1 |
TC0900007010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |