Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017289.hg.1 |
TC01001096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017289.hg.1 |
ENST00000355594.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017289.hg.1 |
ENST00000453670.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017289.hg.1 |
ENST00000544626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017289.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017291.hg.1 |
TC01001096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017291.hg.1 |
ENST00000355594.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017291.hg.1 |
ENST00000544626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017291.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216537.hg.1 |
TC0100015593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216536.hg.1 |
TC0100015593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216535.hg.1 |
TC0100015593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100216533.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216533.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216533.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216533.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216533.hg.1 |
TC0100015593.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000355594.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
ENST00000544626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_001280799.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015593.hg.1 |
NM_144698.4 |
Refseq |