Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000282479.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
ENST00000339673.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_001079911.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008084.hg.1 |
NM_004407.3 |
Refseq |