Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027857.hg.1 |
TC01001781.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027857.hg.1 |
ENST00000366971.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027857.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027854.hg.1 |
TC01001781.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027854.hg.1 |
ENST00000366971.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027854.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027854.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027855.hg.1 |
TC01001781.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027855.hg.1 |
ENST00000366971.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027855.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027855.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185066.hg.1 |
TC0100011549.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185068.hg.1 |
TC0100011549.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000474693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
ENST00000579295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011549.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
ENST00000419102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
ENST00000483790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185067.hg.1 |
TC0100011549.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185070.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185070.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185070.hg.1 |
ENST00000366971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185070.hg.1 |
NM_014053.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185070.hg.1 |
TC0100011549.hg.1 |
Clariom D TC |