Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308018.1 | NP_001294947.1 | 10 - 11 | 1184 | 101 | |
NM_018712.3 | NP_061182.3 | 9 - 10 | 1048 | 101 | ||
GenBank mRNA | AK297351.1 | - | 10 - 11 | 1184 | 101 | |
AL359601.1 | - | 3 - 4 | 235 | 101 | ||
BC028725.2 | - | 9 - 10 | 791 | 4815682 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |