Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010763.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010763.hg.1 |
ENST00000368994.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010763.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010763.hg.1 |
TC10000884.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000507495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
ENST00000507495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162636.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162636.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162636.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162636.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162636.hg.1 |
TC1000009145.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000260723.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000495370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000507495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
ENST00000507495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009145.hg.1 |
NM_144587.3 |
Refseq |