Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027815.hg.1 |
TC05001997.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027815.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027815.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188218.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188218.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188218.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188218.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188218.hg.1 |
TC0500012646.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
ENST00000231228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012646.hg.1 |
NM_002187.2 |
Refseq |