Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007388.hg.1 |
TC06000613.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007388.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007388.hg.1 |
ENST00000542245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007388.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007390.hg.1 |
TC06000613.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007390.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007390.hg.1 |
ENST00000542245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007390.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000524554.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
ENST00000533604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008122.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
ENST00000398776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
ENST00000526118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
ENST00000532171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
NM_007058.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156800.hg.1 |
TC0600008122.hg.1 |
Clariom D TC |