Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000092.4 | NP_000083.3 | 45 - 46 | 4990 | 73 | |
XM_005246281.3 | XP_005246338.1 | 45 - 46 | 4524 | 73 | ||
XM_005246282.3 | XP_005246339.1 | 39 - 40 | 4107 | 73 | ||
XM_011510557.2 | XP_011508859.1 | 46 - 47 | 4581 | 73 | ||
XM_011510558.2 | XP_011508860.1 | 44 - 45 | 4554 | 73 | ||
XM_011510559.2 | XP_011508861.1 | 45 - 46 | 4662 | 73 | ||
XM_011510560.2 | XP_011508862.1 | 45 - 46 | 4662 | 73 | ||
XM_011510561.2 | XP_011508863.1 | 45 - 46 | 4662 | 73 | ||
XM_011510562.2 | XP_011508864.1 | 45 - 46 | 4662 | 73 | ||
XM_011510572.2 | XP_011508874.1 | 24 - 25 | 2887 | 73 | ||
XM_017003296.1 | XP_016858785.1 | 45 - 46 | 4662 | 73 | ||
XM_017003297.1 | XP_016858786.1 | 45 - 46 | 4545 | 73 | ||
XM_017003298.1 | XP_016858787.1 | 45 - 46 | 4662 | 73 | ||
XM_017003299.1 | XP_016858788.1 | 45 - 46 | 4662 | 73 | ||
GenBank mRNA | D17391.1 | - | 7 - 8 | 683 | 73 | |
X81053.1 | - | 45 - 46 | 4544 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons |