Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004819.2 | NP_004810.2 | 18 - 19 | 2737 | 93 | |
XM_005259286.1 | XP_005259343.1 | 18 - 19 | 2690 | 93 | ||
XM_011527354.1 | XP_011525656.1 | 19 - 20 | 2740 | 93 | ||
GenBank mRNA | AB209024.1 | - | 18 - 19 | 4900 | 93 | |
AB210010.1 | - | 18 - 19 | 2582 | 93 | ||
AK130602.1 | - | 9 - 10 | 876 | 93 | ||
AK300252.1 | - | 18 - 19 | 2591 | 93 | ||
BC006536.2 | - | 17 - 18 | 2444 | 93 | ||
BC006567.2 | - | 17 - 18 | 2444 | 93 | ||
BX649165.1 | - | 17 - 18 | 3643 | 93 | ||
U49240.1 | - | 13 - 14 | 2206 | 93 | ||
Y10931.1 | - | 18 - 19 | 2555 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |