Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015346.3 | NP_056161.2 | 31 - 32 | 5935 | 63 | |
XM_017021124.1 | XP_016876613.1 | 31 - 32 | 5954 | 63 | ||
XM_017021125.1 | XP_016876614.1 | 31 - 32 | 5954 | 63 | ||
XM_017021126.1 | XP_016876615.1 | 30 - 31 | 6568 | 63 | ||
XM_017021127.1 | XP_016876616.1 | 20 - 21 | 3650 | 63 | ||
XM_017021128.1 | XP_016876617.1 | 20 - 21 | 3503 | 63 | ||
GenBank mRNA | AB002319.1 | - | 16 - 17 | 2805 | 63 | |
AB425197.1 | - | 31 - 32 | 5923 | 63 | ||
AK024743.1 | - | 3 - 4 | 297 | 63 | ||
BC144594.1 | - | 29 - 30 | 5239 | 63 | ||
BX537886.1 | - | 31 - 32 | 5929 | 63 | ||
BX538025.1 | - | 27 - 28 | 5428 | 63 | ||
BX648683.1 | - | 31 - 32 | 5933 | 63 | ||
CR749276.1 | - | 32 - 33 | 5870 | 63 | ||
CR749365.1 | - | 31 - 32 | 5934 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |