Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027885.hg.1 |
TC06002235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027885.hg.1 |
ENST00000336451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027885.hg.1 |
ENST00000367299.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027885.hg.1 |
ENST00000367303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027885.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027884.hg.1 |
TC06002235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027884.hg.1 |
ENST00000336451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027884.hg.1 |
ENST00000367299.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027884.hg.1 |
ENST00000367303.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027884.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193670.hg.1 |
TC0600013574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000491268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013574.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000336451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000367303.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000444024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
ENST00000622845.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
NM_001271937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
NM_017909.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193671.hg.1 |
TC0600013574.hg.1 |
Clariom D TC |