Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021163.hg.1 |
ENST00000221233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021163.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021163.hg.1 |
TC19001555.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
TC1900010746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188844.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000221233.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000593771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000596905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
ENST00000602129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010746.hg.1 |
NM_020158.3 |
Refseq |