Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043045.hg.1 |
TC01002754.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043045.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043045.hg.1 |
ENST00000506472.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043045.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208375.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208375.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208375.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208375.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208375.hg.1 |
TC0100014505.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
ENST00000235345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014505.hg.1 |
NM_015139.2 |
Refseq |