Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000263559.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000373382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000395098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000467852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000489794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000490696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
ENST00000497564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_001035260.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007888.hg.1 |
NM_004896.4 |
Refseq |