Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014727.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014727.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014727.hg.1 |
TC19001077.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014726.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014726.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014726.hg.1 |
TC19001077.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
TC1900009388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175050.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009388.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
TC1900009388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
ENST00000590789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175045.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
TC1900009388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175049.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
NR_027064.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
TC1900009388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
ENST00000448587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
ENST00000587632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175048.hg.1 |
NR_027064.3 |
Refseq |