Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000466201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000466201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013328.hg.1 |
ENST00000378143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013328.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013328.hg.1 |
ENST00000378165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013328.hg.1 |
ENST00000441445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013328.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013328.hg.1 |
TC10001061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013331.hg.1 |
ENST00000378165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013331.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013331.hg.1 |
TC10001061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
ENST00000378143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
ENST00000378165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
ENST00000441445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
ENST00000535341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
ENST00000540259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013319.hg.1 |
TC10001061.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
ENST00000378143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
ENST00000378165.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
ENST00000441445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
ENST00000535341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
ENST00000540259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013324.hg.1 |
TC10001061.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167494.hg.1 |
TC1000009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167498.hg.1 |
TC1000009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167501.hg.1 |
TC1000009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000466201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000466201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000478580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
ENST00000486786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009880.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
ENST00000378150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
ENST00000378165.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
NM_001308295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
NM_004808.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167497.hg.1 |
TC1000009880.hg.1 |
Clariom D TC |