Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021458.hg.1 |
TC04001513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021458.hg.1 |
ENST00000334383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021458.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021458.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021457.hg.1 |
TC04001513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021457.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021457.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000384419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180721.hg.1 |
TC0400011738.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000334383.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000384419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
ENST00000512235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011738.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
ENST00000394427.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
ENST00000507331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
NM_198179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180720.hg.1 |
TC0400011738.hg.1 |
Clariom D TC |